Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp3ccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080594.2 | NP_001074063.2 | 2 - 3 | 564 | 85 | |
XM_005155313.4 | XP_005155370.1 | 2 - 3 | 595 | 85 | ||
XM_005155314.4 | XP_005155371.1 | 2 - 3 | 595 | 85 | ||
GenBank mRNA | BC129382.1 | - | 2 - 3 | 555 | 85 | |
BC165627.1 | - | 2 - 3 | 285 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |