Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329874.1 | NP_001316803.1 | 4 - 5 | 378 | 89 | |
XM_005161207.4 | XP_005161264.1 | 4 - 5 | 364 | 89 | ||
XM_021468917.1 | XP_021324592.1 | 4 - 5 | 346 | 89 | ||
XM_021468918.1 | XP_021324593.1 | 4 - 5 | 346 | 89 | ||
XM_021468919.1 | XP_021324594.1 | 4 - 5 | 346 | 89 | ||
GenBank mRNA | BC129413.1 | - | 4 - 5 | 340 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |