Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329874.1 | NP_001316803.1 | 7 - 8 | 734 | 64 | |
XM_005161207.4 | XP_005161264.1 | 7 - 8 | 720 | 64 | ||
XM_021468917.1 | XP_021324592.1 | 7 - 8 | 702 | 64 | ||
XM_021468918.1 | XP_021324593.1 | 7 - 8 | 702 | 64 | ||
XM_021468919.1 | XP_021324594.1 | 7 - 8 | 702 | 64 | ||
GenBank mRNA | BC129413.1 | - | 7 - 8 | 696 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |