Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: creg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320205.1 | NP_001307134.1 | 1002 | 95 | ||
GenBank mRNA | BC129190.1 | - | 700 | 95 | ||
CF265272.1 | - | 627 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio |