Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: defbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081555.1 | NP_001075024.1 | 1 - 2 | 79 | 67 | |
GenBank mRNA | AM181360.1 | - | 1 - 2 | 79 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |