Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx3xa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126423.1 | NP_001119895.1 | 4 - 5 | 618 | 68 | |
XM_005167718.4 | XP_005167775.1 | 5 - 6 | 722 | 68 | ||
XM_005167720.4 | XP_005167777.1 | 3 - 4 | 614 | 68 | ||
XM_009304751.3 | XP_009303026.1 | 4 - 5 | 662 | 68 | ||
XM_017357705.2 | XP_017213194.1 | 5 - 6 | 719 | 68 | ||
XM_017357706.2 | XP_017213195.1 | 5 - 6 | 722 | 68 | ||
XM_021478726.1 | XP_021334401.1 | 7 - 8 | 1059 | 68 | ||
XM_021478727.1 | XP_021334402.1 | 7 - 8 | 1056 | 68 | ||
XM_021478728.1 | XP_021334403.1 | 7 - 8 | 1059 | 68 | ||
XM_021478729.1 | XP_021334404.1 | 6 - 7 | 1011 | 68 | ||
XM_021478730.1 | XP_021334405.1 | 6 - 7 | 999 | 68 | ||
XM_021478731.1 | XP_021334406.1 | 6 - 7 | 975 | 68 | ||
XM_021478733.1 | XP_021334408.1 | 8 - 9 | 972 | 68 | ||
XM_021478734.1 | XP_021334409.1 | 7 - 8 | 912 | 68 | ||
XM_021478735.1 | XP_021334410.1 | 7 - 8 | 888 | 68 | ||
XM_021478736.1 | XP_021334411.1 | 4 - 5 | 674 | 68 | ||
GenBank mRNA | BC133162.1 | - | 4 - 5 | 618 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |