Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081692.2 | NP_001075161.2 | 2 - 3 | 381 | 62 | |
XM_005160248.4 | XP_005160305.1 | 2 - 3 | 339 | 62 | ||
XM_009293357.3 | XP_009291632.1 | 2 - 3 | 967 | 62 | ||
XM_009293362.3 | XP_009291637.1 | 2 - 3 | 338 | 62 | ||
XM_009293365.3 | XP_009291640.1 | 2 - 3 | 268 | 62 | ||
GenBank mRNA | BC129321.1 | - | 2 - 3 | 383 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |