Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atpaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082821.1 | NP_001076290.1 | 1 - 2 | 380 | 104 | |
GenBank mRNA | BC134167.1 | - | 1 - 2 | 380 | 104 | |
BC153513.1 | - | 1 - 2 | 390 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |