Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083002.1 | NP_001076471.1 | 3 - 4 | 838 | 98 | |
XM_005163128.4 | XP_005163185.1 | 3 - 4 | 1030 | 98 | ||
XM_009298079.3 | XP_009296354.1 | 3 - 4 | 821 | 98 | ||
GenBank mRNA | BC131867.1 | - | 3 - 4 | 838 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |