Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083068.1 | NP_001076537.1 | 3 - 4 | 883 | 91 | |
XM_005159560.3 | XP_005159617.1 | 3 - 4 | 1128 | 91 | ||
XM_009294101.1 | XP_009292376.1 | 2 - 3 | 194 | 91 | ||
GenBank mRNA | BC161621.1 | - | 3 - 4 | 1120 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |