Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacng7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083084.1 | NP_001076553.1 | 4 - 5 | 569 | 74 | |
XM_005159404.4 | XP_005159461.1 | 5 - 6 | 898 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |