Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suv39h1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126482.1 | NP_001119954.1 | 2 - 3 | 608 | 131 | |
GenBank mRNA | AI416110.1 | - | 2 - 3 | 215 | 131 | |
BC052225.1 | - | 2 - 3 | 220 | 131 | ||
BC115190.1 | - | 2 - 3 | 215 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |