Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anpepb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089325.1 | NP_001082794.1 | 6 - 7 | 1305 | 102 | |
GenBank mRNA | BC115260.1 | - | 6 - 7 | 1305 | 102 | |
BC154244.1 | - | 6 - 7 | 1306 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |