Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089553.1 | NP_001083022.1 | 16 - 17 | 2560 | 70 | |
XM_005163851.4 | XP_005163908.1 | 16 - 17 | 2771 | 70 | ||
XM_021468997.1 | XP_021324672.1 | 16 - 17 | 2841 | 70 | ||
XM_021469000.1 | XP_021324675.1 | 16 - 17 | 2823 | 70 | ||
GenBank mRNA | BC139664.1 | - | 16 - 17 | 2560 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |