Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp4cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111168.1 | NP_001104638.1 | 8 - 9 | 1053 | 96 | |
XM_009306399.3 | XP_009304674.1 | 8 - 9 | 1063 | 96 | ||
GenBank mRNA | BC155609.1 | - | 8 - 9 | 1053 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |