Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: march8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009307237.3 | XP_009305512.1 | 1 - 2 | 426 | 79 | |
XM_009307239.3 | XP_009305514.1 | 1 - 2 | 403 | 79 | ||
GenBank mRNA | BC153940.1 | - | 1 - 2 | 215 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |