Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp27c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113337.3 | NP_001106808.2 | 3 - 4 | 805 | 84 | |
GenBank mRNA | BC154632.1 | - | 3 - 4 | 714 | 84 | |
BC171578.1 | - | 3 - 4 | 490 | 84 | ||
EH591543.1 | - | 3 - 4 | 794 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |