Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113501.1 | NP_001106973.1 | 25 - 26 | 3387 | 80 | |
XM_005157694.4 | XP_005157751.1 | 17 - 18 | 2150 | 80 | ||
XM_017350981.2 | XP_017206470.1 | 24 - 25 | 4442 | 80 | ||
GenBank mRNA | AM749802.1 | - | 25 - 26 | 3387 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |