Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113501.1 | NP_001106973.1 | 3 - 4 | 354 | 107 | |
XM_009291655.3 | XP_009289930.1 | 2 - 3 | 107 | |||
XM_017350981.2 | XP_017206470.1 | 2 - 3 | 107 | |||
GenBank mRNA | AM749802.1 | - | 3 - 4 | 354 | 107 | |
EU332350.1 | - | 3 - 4 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |