Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacnb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113519.2 | NP_001106991.1 | 13 - 14 | 1422 | 65 | |
XM_005167837.4 | XP_005167894.1 | 12 - 13 | 1490 | 65 | ||
GenBank mRNA | BC171593.1 | - | 13 - 14 | 1328 | 65 | |
BC171597.1 | - | 13 - 14 | 1328 | 65 | ||
DQ372948.1 | - | 12 - 13 | 1298 | 65 | ||
EF601060.1 | - | 12 - 13 | 1435 | 65 | ||
EF601061.1 | - | 13 - 14 | 1426 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |