Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113524.1 | NP_001106996.1 | 4 - 5 | 586 | 119 | |
XM_005166063.4 | XP_005166120.1 | 4 - 5 | 1442 | 119 | ||
GenBank mRNA | EU099311.1 | - | 4 - 5 | 586 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |