Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113578.1 | NP_001107050.1 | 10 - 11 | 1264 | 110 | |
GenBank mRNA | BC154609.1 | - | 10 - 11 | 1264 | 110 | |
BC171427.1 | - | 10 - 11 | 1264 | 110 | ||
BC171429.1 | - | 10 - 11 | 1264 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |