Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113617.2 | NP_001107089.2 | 1 - 2 | 112 | 69 | |
GenBank mRNA | BC154809.1 | - | 1 - 2 | 71 | 69 | |
EH486004.1 | - | 1 - 2 | 104 | 69 | ||
EH580492.1 | - | 1 - 2 | 138 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |