Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113621.1 | NP_001107093.1 | 7 - 8 | 1369 | 70 | |
XM_017358910.2 | XP_017214399.1 | 7 - 8 | 2142 | 70 | ||
XM_017358916.2 | XP_017214405.1 | 5 - 6 | 733 | 70 | ||
XM_017358919.2 | XP_017214408.1 | 7 - 8 | 2161 | 70 | ||
GenBank mRNA | BC154444.1 | - | 7 - 8 | 1369 | 70 | |
BC171521.1 | - | 7 - 8 | 1364 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |