Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113621.1 | NP_001107093.1 | 9 - 10 | 1717 | 73 | |
XM_017358910.2 | XP_017214399.1 | 9 - 10 | 2490 | 73 | ||
XM_017358916.2 | XP_017214405.1 | 7 - 8 | 1081 | 73 | ||
XM_017358919.2 | XP_017214408.1 | 9 - 10 | 2509 | 73 | ||
GenBank mRNA | BC154444.1 | - | 9 - 10 | 1717 | 73 | |
BC171521.1 | - | 9 - 10 | 1712 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |