Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkain4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113646.1 | NP_001107118.1 | 4 - 5 | 698 | 56 | |
XM_005161981.4 | XP_005162038.1 | 4 - 5 | 887 | 56 | ||
GenBank mRNA | BC154589.1 | - | 4 - 5 | 698 | 56 | |
BC171601.1 | - | 4 - 5 | 598 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |