Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114406.1 | NP_001107878.1 | 4 - 5 | 704 | 65 | |
XM_005169892.3 | XP_005169949.1 | 4 - 5 | 756 | 65 | ||
GenBank mRNA | BC068411.1 | - | 3 - 4 | 270 | 65 | |
BC159253.1 | - | 4 - 5 | 704 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |