Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114407.2 | NP_001107879.2 | 6 - 7 | 615 | 70 | |
GenBank mRNA | BC155085.1 | - | 6 - 7 | 615 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |