Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt5g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114442.1 | NP_001107914.1 | 3 - 4 | 281 | 109 | |
XM_021477582.1 | XP_021333257.1 | 2 - 3 | 254 | 109 | ||
GenBank mRNA | BC159244.1 | - | 3 - 4 | 281 | 109 | |
GU299167.1 | - | 3 - 4 | 242 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |