Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130982.1 | NP_571057.1 | 1 | 446 | 90 | |
GenBank mRNA | AF084949.1 | - | 1 | 446 | 90 | |
BC162864.1 | - | 1 | 548 | 90 | ||
BC162904.1 | - | 1 | 548 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |