Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tle3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313694.1 | NP_001300623.1 | 15 - 16 | 1691 | 70 | |
NM_131012.2 | NP_571087.2 | 15 - 16 | 1688 | 70 | ||
XM_005159039.4 | XP_005159096.1 | 16 - 17 | 2076 | 70 | ||
XM_005159041.4 | XP_005159098.1 | 16 - 17 | 2073 | 70 | ||
XM_017351898.2 | XP_017207387.1 | 16 - 17 | 2073 | 70 | ||
GenBank mRNA | BC162533.1 | - | 15 - 16 | 1589 | 70 | |
U96340.1 | - | 15 - 16 | 1591 | 70 | ||
Y12466.1 | - | 1591 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |