Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ihhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131088.1 | NP_571163.1 | 2 - 3 | 558 | 85 | |
GenBank mRNA | Y08426.1 | - | 558 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |