Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ldb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001362321.1 | NP_001349250.1 | 9 - 10 | 957 | 68 | |
NM_131313.3 | NP_571388.3 | 9 - 10 | 1092 | 68 | ||
GenBank mRNA | AF031375.1 | - | 9 - 10 | 1075 | 68 | |
BC078225.1 | - | 9 - 10 | 1093 | 68 | ||
BC093137.1 | - | 9 - 10 | 957 | 68 | ||
BC110113.1 | - | 9 - 10 | 1010 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |