Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: neurog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131815.1 | NP_571890.1 | 1 - 2 | 63 | 75 | |
GenBank mRNA | AF181996.1 | - | 1 - 2 | 63 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Inventoried Probe spans exons |