Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mitfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131847.2 | NP_571922.2 | 5 - 6 | 853 | 69 | |
XM_021469718.1 | XP_021325393.1 | 4 - 5 | 879 | 69 | ||
GenBank mRNA | JN105118.1 | - | 5 - 6 | 853 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |