Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153666.2 | NP_705952.2 | 6 - 7 | 945 | 85 | |
GenBank mRNA | AB036516.1 | - | 6 - 7 | 945 | 85 | |
BC163297.1 | - | 6 - 7 | 833 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |