Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173247.1 | NP_775354.1 | 2 - 3 | 664 | 132 | |
GenBank mRNA | AF506216.1 | - | 2 - 3 | 664 | 132 | |
AY398420.1 | - | 2 - 3 | 667 | 132 | ||
BC059462.1 | - | 2 - 3 | 667 | 132 | ||
BC065434.1 | - | 2 - 3 | 661 | 132 | ||
BC085562.1 | - | 1621 | 132 | |||
BC152234.1 | - | 3 - 4 | 927 | 132 | ||
BC153470.1 | - | 2 - 3 | 701 | 132 | ||
BC154238.1 | - | 1193 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |