Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178219.2 | NP_835737.1 | 2 - 3 | 410 | 61 | |
GenBank mRNA | AY182171.1 | - | 2 - 3 | 410 | 61 | |
BC162282.1 | - | 2 - 3 | 361 | 61 | ||
BC162831.1 | - | 2 - 3 | 361 | 61 | ||
KM655731.1 | - | 2 - 3 | 338 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |