Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: heyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181736.1 | NP_859425.1 | 3 - 4 | 278 | 77 | |
GenBank mRNA | AJ510222.1 | - | 3 - 4 | 278 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |