Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hbae1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182940.3 | NP_891985.1 | 1 - 2 | 132 | 87 | |
GenBank mRNA | AY325264.1 | - | 1 - 2 | 100 | 87 | |
BC071550.1 | - | 1 - 2 | 132 | 87 |
Gene Symbol: hbae1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365350.1 | NP_001352279.1 | 1 - 2 | 135 | 87 | |
XM_001333519.7 | XP_001333555.1 | 1 - 2 | 164 | 87 | ||
GenBank mRNA | BC154182.1 | - | 1 - 2 | 132 | 87 |
Gene Symbol: hbae1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365351.1 | NP_001352280.1 | 1 - 2 | 135 | 87 | |
GenBank mRNA | BC152154.1 | - | 1 - 2 | 132 | 87 | |
BC155770.1 | - | 1 - 2 | 132 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |