Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnf1bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194372.3 | NP_919353.1 | 7 - 8 | 1506 | 94 | |
GenBank mRNA | AF250352.1 | - | 7 - 8 | 1506 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |