Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194404.1 | NP_919385.1 | 6 - 7 | 813 | 72 | |
GenBank mRNA | AF286179.1 | - | 6 - 7 | 813 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |