Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc15a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198064.1 | NP_932330.1 | 17 - 18 | 1531 | 78 | |
GenBank mRNA | AY300011.1 | - | 17 - 18 | 1531 | 78 | |
BC054899.1 | - | 17 - 18 | 1531 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |