Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198808.1 | NP_942103.1 | 9 - 10 | 930 | 91 | |
XM_021478992.1 | XP_021334667.1 | 8 - 9 | 1237 | 91 | ||
GenBank mRNA | BC054569.1 | - | 9 - 10 | 930 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |