Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198815.2 | NP_942110.2 | 4 - 5 | 783 | 70 | |
GenBank mRNA | AY217090.1 | - | 3 - 4 | 587 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |