Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: depdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198872.1 | NP_942573.1 | 8 - 9 | 1950 | 60 | |
GenBank mRNA | BC044382.1 | - | 8 - 9 | 1950 | 60 | |
BC171596.1 | - | 8 - 9 | 1902 | 60 | ||
BC171624.1 | - | 8 - 9 | 1902 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |