Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199796.1 | NP_956090.1 | 2 - 3 | 283 | 131 | |
XM_005166011.4 | XP_005166068.1 | 1 - 2 | 405 | 131 | ||
GenBank mRNA | BC044473.1 | - | 2 - 3 | 283 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |