Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199958.1 | NP_956252.1 | 5 - 6 | 807 | 69 | |
XM_021467596.1 | XP_021323271.1 | 5 - 6 | 69 | |||
GenBank mRNA | BC054587.1 | - | 5 - 6 | 807 | 69 | |
BC163161.1 | - | 5 - 6 | 69 | |||
BC163187.1 | - | 5 - 6 | 69 | |||
BC164454.1 | - | 4 - 5 | 579 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |