Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328575.1 | NP_001315504.1 | 3 - 4 | 393 | 80 | |
NM_001328576.1 | NP_001315505.1 | 3 - 4 | 350 | 80 | ||
NM_200021.2 | NP_956315.1 | 3 - 4 | 396 | 80 | ||
GenBank mRNA | BC045369.1 | - | 3 - 4 | 397 | 80 | |
BC067598.1 | - | 2 - 3 | 327 | 80 | ||
BC165424.1 | - | 2 - 3 | 340 | 80 | ||
EH491524.1 | - | 3 - 4 | 365 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |