Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200061.2 | NP_956355.2 | 3 - 4 | 279 | 72 | |
GenBank mRNA | BC053181.1 | - | 3 - 4 | 279 | 72 | |
BC164909.1 | - | 3 - 4 | 263 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |